4-(Methylthio)phenylacetic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
M2874-1G
M2874-5G
TCIAM2874-1GEA
375
SEK
TCIAM2874-1G
TCIAM2874-5G
4-(Methylthio)phenylacetic acid ≥98.0% (by GC, titration analysis)
4-(Methylthio)phenylacetic acid
Formel:
C₉H₁₀O₂S MW: 182.24 g/mol |
MDL Number:
MFCD00192325 CAS nummer: 16188-55-9 |
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...