3-(Methylaminomethyl)benzonitrile ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 3-((Methylamino)methyl)benzonitrile

M2869-1G M2869-5G
TCIAM2869-1GEA 910 SEK
TCIAM2869-1G TCIAM2869-5G
3-(Methylaminomethyl)benzonitrile ≥98.0% (by GC, titration analysis)
3-(Methylaminomethyl)benzonitrile
Formel: C₉H₁₀N₂
MW: 146,193 g/mol
Kokpunkt: 145°C
MDL Number: MFCD09731650
CAS nummer: 90389-96-1
UN: 2922
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcca499f996977ff833896cc9364205c3b343609&vid=98939480d66cbdf9419696275965e6cc77b62977&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR