Moc-Val-OH ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: N-(Methoxycarbonyl)-L-valine, MOC-Val-OH

M2857-5G M2857-25G
TCIAM2857-5GEA 230 SEK
TCIAM2857-5G TCIAM2857-25G
Moc-Val-OH ≥98.0% (by HPLC, titration analysis)
Moc-Val-OH
Formel: C₇H₁₃NO₄
MW: 175,2 g/mol
MDL Number: MFCD12795357
CAS nummer: 74761-42-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db502cca1567ba0cd349b178e42742decfb269ef&vid=db502cca1567ba0cd349b178e42742decfb269ef&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 109.0 to 113.0 deg-C
Specific rotation [a]20/D -5.0 to -8.0 deg (C=1,AcOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR