(S)-3-Methylmorpholine hydrochloride ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
M2848-1G
M2848-5G
TCIAM2848-1GEA
1275
SEK
TCIAM2848-1G
TCIAM2848-5G
(S)-3-Methylmorpholine hydrochloride ≥98.0%
(S)-3-Methylmorpholine hydrochloride
Formel:
C₅H₁₁NO·ClH MW: 137,61 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD18382512 CAS nummer: 1022094-03-6 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 125.0 to 130.0 deg-C |
Specific rotation [a]20/D | -2.0 to -5.0 deg (C=1, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...