Methyl 2-Fluoro-2-methylpropionate ≥95.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
2-Fluoro-2-methylpropionic acid methyl ester
M2827-5G
M2827-25G
TCIAM2827-5GEA
885
SEK
TCIAM2827-5G
TCIAM2827-25G
Methyl 2-Fluoro-2-methylpropionate ≥95.0%
Methyl 2-Fluoro-2-methylpropionate
Formel:
C₅H₉FO₂ MW: 120,12 g/mol Kokpunkt: 109°C |
MDL Number:
MFCD03265444 CAS nummer: 338-76-1 UN: 2924 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...