3-Methyl-N-methylbenzylamine ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: N-Methyl-1-(m-tolyl)methanamine

M2824-1ML M2824-5ML
TCIAM2824-1MLEA 685 SEK
TCIAM2824-1ML TCIAM2824-5ML
3-Methyl-N-methylbenzylamine ≥97.0%
3-Methyl-N-methylbenzylamine
Formel: C₉H₁₃N
MW: 135,21 g/mol
Kokpunkt: 92°C
Flampunkt: 78°C
MDL Number: MFCD05865181
CAS nummer: 39180-84-2
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=301605430916d4f0e32339fd00b8d37912e1dc96&vid=53ed6a8e476e4e61f0a2048ad465657379b2f8b8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR