9-Methyl-9H-pyrido[3,4-b]indole ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
9-Methyl-beta-carboline
M2818-1G
M2818-5G
TCIAM2818-1GEA
1625
SEK
TCIAM2818-1G
TCIAM2818-5G
9-Methyl-9H-pyrido[3,4-b]indole ≥98.0% (by GC, titration analysis)
9-Methyl-9H-pyrido[3,4-b]indole
Formel:
C₁₂H₁₀N₂ MW: 182,23 g/mol |
MDL Number:
MFCD18803860 CAS nummer: 2521-07-5 |
Specifikation testresultat
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 105.0 to 109.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...