Methyl-5-amino-2-chlorobenzoate ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Methyl 5-amino-2-chlorobenzoate
M2812-1G M2812-5G
TCIAM2812-1GEA 375 SEK
TCIAM2812-1G TCIAM2812-5G
Methyl-5-amino-2-chlorobenzoate ≥98.0%
Methyl-5-amino-2-chlorobenzoate
Formel: C₈H₈ClNO₂
MW: 185,6100 g/mol
Kokpunkt: 160°C
Storage Temperature: Kylskåp
MDL Number: MFCD07800315
CAS nummer: 42122-75-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da171a6b991185d17400e8e04272c6f3a4c9e215&vid=da171a6b991185d17400e8e04272c6f3a4c9e215&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to lump
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %