Methyl-4-hydroxy-3,5-dimethoxybenzoate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Methyl 4-Hydroxy-3,5-Dimethoxybenzoate, Methyl syringate
M2806-5G
M2806-25G
TCIAM2806-5GEA
510
SEK
TCIAM2806-5G
TCIAM2806-25G
Methyl-4-hydroxy-3,5-dimethoxybenzoate ≥98.0%
Methyl-4-hydroxy-3,5-dimethoxybenzoate
Formel:
C₁₀H₁₂O₅ MW: 212,20 g/mol |
MDL Number:
MFCD00017199 CAS nummer: 884-35-5 EINECS: 429-050-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 106.0 to 110.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...