Methyl oxindole-6-carboxylate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Methyl 2-Oxoindoline-6-carboxylate, 2-Oxoindoline-6-carboxylic acid methyll ester

M2799-1G M2799-5G
TCIAM2799-1GEA 685 SEK
TCIAM2799-1G TCIAM2799-5G
Methyl oxindole-6-carboxylate ≥98.0%
Methyl oxindole-6-carboxylate
Formel: C₁₀H₉NO₃
MW: 191,1800 g/mol
MDL Number: MFCD03095196
CAS nummer: 14192-26-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ae1205a460a2b26c6be6dcd99142be0bfa8c496&vid=7ae1205a460a2b26c6be6dcd99142be0bfa8c496&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 210.0 to 214.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR