3,3'-Methylenebis[1-(2,6-diisopropylphenyl)-3-imidazolium bromide] ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

M2791-1G M2791-5G
TCIAM2791-1GEA 2150 SEK
TCIAM2791-1G TCIAM2791-5G
3,3'-Methylenebis[1-(2,6-diisopropylphenyl)-3-imidazolium bromide] ≥98.0% (by HPLC, titration analysis)
3,3'-Methylenebis[1-(2,6-diisopropylphenyl)-3-imidazolium bromide]
Formel: C₃₁H₄₂Br₂N₄
MW: 630,51 g/mol
Smältpunkt: 277°C
CAS nummer: 889617-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=045aa8639a272e6b223e17a9f7a2ebfa9128fc64&vid=045aa8639a272e6b223e17a9f7a2ebfa9128fc64&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR