2-(4-Methoxyphenyl)indole ≥98.0%
Leverantör: TCI
M2789-1G
M2789-5G
TCIAM2789-1GEA
1500
SEK
TCIAM2789-1G
TCIAM2789-5G
2-(4-Methoxyphenyl)indole ≥98.0%
2-(4-Methoxyphenyl)indole
Formel:
C₁₅H₁₃NO |
MDL Number:
MFCD00443483 CAS nummer: 5784-95-2 |
|
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Melting point | 229.0 to 233.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...