2-Acetylamino-4-methylpyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: N-(4-Methyl-2-pyridyl)acetamide, N-(4-Methylpyridin-2-yl)acetamide, 2-Acetamido-4-picoline, 2-Acetamido-4-methylpyridine

M2787-1G M2787-5G
TCIAM2787-1GEA 415 SEK
TCIAM2787-1G TCIAM2787-5G
2-Acetylamino-4-methylpyridine ≥98.0%
2-Acetylamino-4-methylpyridine
Formel: C₈H₁₀N₂O
MW: 150,1778 g/mol
MDL Number: MFCD00234432
CAS nummer: 5327-32-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a571fa2323dddf31df087e875a67d7fe84b70885&vid=a571fa2323dddf31df087e875a67d7fe84b70885&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR