1-Methylpyrrole-2-carbonitrile ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1-Methyl-1H-pyrrole-2-carbonitrile, 2-Cyano-1-methylpyrrole
M2784-1G M2784-5G
TCIAM2784-1GEA 1025 SEK
TCIAM2784-1G TCIAM2784-5G
1-Methylpyrrole-2-carbonitrile ≥98.0%
1-Methylpyrrole-2-carbonitrile
Formel: C₆H₆N₂
MW: 106,13 g/mol
Kokpunkt: 87°C
MDL Number: MFCD02693721
CAS nummer: 34884-10-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80a5accc91a556747a1326b778e8f2d37d3bcc6b&vid=80a5accc91a556747a1326b778e8f2d37d3bcc6b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %