1-Methyl-2-(2-pyridyl)pyridinium iodide ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 1-Methyl-2-(2-pyridyl)pyridinium Iodide

M2778-200MG M2778-1G
TCIAM2778-200MGEA 710 SEK
TCIAM2778-200MG TCIAM2778-1G
1-Methyl-2-(2-pyridyl)pyridinium iodide ≥98.0% (by HPLC, titration analysis)
1-Methyl-2-(2-pyridyl)pyridinium iodide
Formel: C₁₁H₁₁IN₂
MW: 298.13 g/mol
MDL Number: MFCD23700876
CAS nummer: 77972-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3731faa12788dcfe91d34f541bc6203fc643ca1&vid=c3731faa12788dcfe91d34f541bc6203fc643ca1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR