1-(4-Methylphenyl)-3-phenylprop-2-en-1-one ≥98.0%

Leverantör: TCI

Synonyms: (E)-2-Benzal-4'-methylacetophenone, (E)-2-Benzylidene-4'-methylacetophenone, (E)-4'-Methylchalcone

M2777-1G M2777-5G
TCIAM2777-1GEA 1125 SEK
TCIAM2777-1G TCIAM2777-5G
1-(4-Methylphenyl)-3-phenylprop-2-en-1-one ≥98.0%
1-(4-Methylphenyl)-3-phenylprop-2-en-1-one
Formel: C₁₆H₁₄O
MDL Number: MFCD00027612
CAS nummer: 14802-30-3

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR