4-(1-Propynyl)phthalic anhydride ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 4-(Methylethynyl)phthalic anhydride

M2757-5G M2757-25G
TCIAM2757-5GEA 1025 SEK
TCIAM2757-5G TCIAM2757-25G
4-(1-Propynyl)phthalic anhydride ≥98.0% (by GC, titration analysis)
4-(1-Propynyl)phthalic anhydride
Formel: C₁₁H₆O₃
CAS nummer: 1240685-26-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1aff12f9acc66046180d439bc97c1157e113410f&vid=1aff12f9acc66046180d439bc97c1157e113410f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Morpholine Method) min. 98.0 %
Melting point 113.0 to 117.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR