4-Acetyl-4'-methoxydiphenyl ether ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1-(4-(4-Methoxyphenoxy)phenyl)ethanone, 4'-(4-Methoxyphenoxy)acetophenone, 4-Acetyl-4'-methoxydiphenyl ether

M2740-1G M2740-5G
TCIAM2740-1GEA 855 SEK
TCIAM2740-1G TCIAM2740-5G
4-Acetyl-4'-methoxydiphenyl ether ≥98.0%
4-Acetyl-4'-methoxydiphenyl ether
Formel: C₁₅H₁₄O₃
MW: 242,28 g/mol
Kokpunkt: 236°C
MDL Number: MFCD00052928
CAS nummer: 54916-28-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4a8e3cefc8ddab1a4211d7e2e6e26cd51098cc7&vid=b4a8e3cefc8ddab1a4211d7e2e6e26cd51098cc7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR