1-Methyl-3-octylimidazolium tetrafluoroborate ≥97.0% (by HPLC, total nitrogen)

Leverantör: TCI
Varning

M2732-25G M2732-5G
TCIAM2732-25GEA 1425 SEK
TCIAM2732-25G TCIAM2732-5G
1-Methyl-3-octylimidazolium tetrafluoroborate ≥97.0% (by HPLC, total nitrogen)
1-Methyl-3-octylimidazolium tetrafluoroborate
Formel: C₁₂H₂₃BF₄N₂
MDL Number: MFCD03095434
CAS nummer: 244193-52-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5d2b6c37f8caadbb8179f07ec3dd74ac85ff177&vid=a5d2b6c37f8caadbb8179f07ec3dd74ac85ff177&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR