Methyl-6-fluoronicotinate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 6-Fluoronicotinic acid methyl ester, Methyl 6-fluoropyridine-3-carboxylate

M2724-1G M2724-5G
TCIAM2724-1GEA 455 SEK
TCIAM2724-1G TCIAM2724-5G
Methyl-6-fluoronicotinate ≥98.0%
Methyl-6-fluoronicotinate
Formel: C₇H₆FNO₂
MW: 155,13 g/mol
MDL Number: MFCD03095098
CAS nummer: 1427-06-1

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 49.0 to 53.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR