tert-Butyl hydrogen malonate ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 3-Tert-butoxy-3-oxopropanoic acid, mono-tert-Butyl malonate
M2687-5G M2687-25G
TCIAM2687-5GEA 390 SEK
TCIAM2687-5G TCIAM2687-25G
tert-Butyl hydrogen malonate ≥97.0% (by GC, titration analysis)
tert-Butyl hydrogen malonate
Formel: C₇H₁₂O₄
MW: 160,17 g/mol
Kokpunkt: 90°C
Smältpunkt: 19°C
Flampunkt: 91°C
MDL Number: MFCD00191886
CAS nummer: 40052-13-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a186838c005190742420395ba26d1e63d4fda0cd&vid=a186838c005190742420395ba26d1e63d4fda0cd&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %