N-Methylpyridin-3-amine ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
Synonyms: 3-(Methylamino)pyridine
M2684-1G M2684-5G
TCIAM2684-1GEA 445 SEK
TCIAM2684-1G TCIAM2684-5G
N-Methylpyridin-3-amine ≥98.0% (by GC, titration analysis)
N-Methylpyridin-3-amine
Formel: C₆H₈N₂
MW: 108.144 g/mol
Kokpunkt: 110°C
MDL Number: MFCD01418130
CAS nummer: 18364-47-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c614415673f63be85bf2009ad0650a54290c428&vid=9c614415673f63be85bf2009ad0650a54290c428&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %