4-(Methoxymethyl)benzeneboronic acid (contains varying amounts of Anhydride)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-(Methoxymethyl)phenylboronic acid, (4-(Methoxymethyl)phenyl)boronic acid
M2677-1G
M2677-5G
TCIAM2677-1GEA
695
SEK
TCIAM2677-1G
TCIAM2677-5G
4-(Methoxymethyl)benzeneboronic acid (contains varying amounts of Anhydride)
4-(Methoxymethyl)benzeneboronic acid
Formel:
C₈H₁₁BO₃ MW: 165,983 g/mol |
MDL Number:
MFCD03788426 CAS nummer: 279262-11-2 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...