2-Morpholinoaniline ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2-(4-Morpholinyl)aniline

M2671-5G M2671-25G
TCIAM2671-5GEA 510 SEK
TCIAM2671-5G TCIAM2671-25G
2-Morpholinoaniline ≥98.0% (by GC, titration analysis)
2-Morpholinoaniline
Formel: C₁₀H₁₄N₂O
MW: 178,23 g/mol
Kokpunkt: 120°C
MDL Number: MFCD00047408
CAS nummer: 5585-33-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=978e8f2d136a184e2a7e25d6ef7bc7670b940d54&vid=978e8f2d136a184e2a7e25d6ef7bc7670b940d54&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 94.0 to 98.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR