2-Methoxy-4-methyl-5-nitropyridine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
M2652-1G
M2652-5G
TCIAM2652-1GEA
415
SEK
TCIAM2652-1G
TCIAM2652-5G
2-Methoxy-4-methyl-5-nitropyridine ≥98.0%
2-Methoxy-4-methyl-5-nitropyridine
Formel:
C₇H₈N₂O₃ MW: 168,15 g/mol |
MDL Number:
MFCD03095075 CAS nummer: 6635-90-1 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 79.0 to 83.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...