4-Methoxybenzenediazonium tetrafluoroborate ≥98.0%

Leverantör: TCI EUROPE
Fara

M2636-5G M2636-25G
TCIAM2636-5GEA 1325 SEK
TCIAM2636-5G TCIAM2636-25G
4-Methoxybenzenediazonium tetrafluoroborate ≥98.0%
4-Methoxybenzenediazonium tetrafluoroborate
Formel: C₇H₇BF₄N₂O
MW: 221,95 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00011897
CAS nummer: 459-64-3
EINECS: 207-296-2
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa40153b7669cff2fa71c2599674ad6a1d57bb14&vid=fa40153b7669cff2fa71c2599674ad6a1d57bb14&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR