2-Methylquinoline-6-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
M2625-5G
M2625-25G
TCIAM2625-5GEA
495
SEK
TCIAM2625-5G
TCIAM2625-25G
2-Methylquinoline-6-carboxylic acid ≥98.0% (by HPLC, titration analysis)
2-Methylquinoline-6-carboxylic acid
Formel:
C₁₁H₉NO₂ MW: 187,19 g/mol |
MDL Number:
MFCD00748504 CAS nummer: 635-80-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...