Methyl-2,6-dichloronicotinate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Methyl 2,6-dichloronicotinate

M2616-1G M2616-5G
TCIAM2616-1GEA 270 SEK
TCIAM2616-1G TCIAM2616-5G
Methyl-2,6-dichloronicotinate ≥98.0%
Methyl-2,6-dichloronicotinate
Formel: C₇H₅Cl₂NO₂
MW: 206,03 g/mol
MDL Number: MFCD07369794
CAS nummer: 65515-28-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc3073f293ef6168feba0f1e17840cb71c8b6a12&vid=bc3073f293ef6168feba0f1e17840cb71c8b6a12&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR