1-(2-Methyl-5-nitrophenyl)guanidine nitrate ≥97.0% (by HPLC, titration analysis)

Leverantör: TCI
Varning

M2613-1G M2613-5G
TCIAM2613-1GEA 910 SEK
TCIAM2613-1G TCIAM2613-5G
1-(2-Methyl-5-nitrophenyl)guanidine nitrate ≥97.0% (by HPLC, titration analysis)
1-(2-Methyl-5-nitrophenyl)guanidine nitrate
Formel: C₈H₁₀N₄O₂
Smältpunkt: 218°C
MDL Number: MFCD09842424
CAS nummer: 152460-08-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c5950dba29ac3af8695b0d0422b70a828b2fa01&vid=0c5950dba29ac3af8695b0d0422b70a828b2fa01&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR