Toluen-α-sulfonylfluorid ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulphonyl fluoride
B22146.06 B22146.03 B22146.14
B22146.06EA 1950 SEK
B22146.06 B22146.03 B22146.14
Toluen-α-sulfonylfluorid ≥99%
Toluen-α-sulfonylfluorid
Purity: 99% CAS No.: 329-98-6 Molecular Formula: C7H7FO2S Formula Weight: 174.19 Melting Point: 92-94° Beilstein Reference: 2088311 UN No.: 2923 MDL No.: MFCD00007424
Formel: C₇H₇FO₂S
MW: 174,2 g/mol
Kokpunkt: 80…85 °C (0,3 mmHg)
Smältpunkt: 92…94 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007424
CAS nummer: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8,III