Methyl-4-bromo-3-nitrobenzoate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Bromobenzoic acid methyl ester, Methyl 4-bromo-3-nitrobenzoate

M2596-1G M2596-5G
TCIAM2596-1GEA 215 SEK
TCIAM2596-1G TCIAM2596-5G
Methyl-4-bromo-3-nitrobenzoate ≥98.0%
Methyl-4-bromo-3-nitrobenzoate
Formel: C₈H₆BrNO₄
MW: 260,04 g/mol
MDL Number: MFCD00553089
CAS nummer: 2363-16-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e15d0a55ae8127e449b31299292770e3fc10f968&vid=e15d0a55ae8127e449b31299292770e3fc10f968&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 101.0 to 105.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR