Methyl-3-bromothiophene-2-carboxylate ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Methyl 3-bromothiophene-2-carboxylate
M2590-1G
M2590-5G
TCIAM2590-1GEA
775
SEK
TCIAM2590-1G
TCIAM2590-5G
Methyl-3-bromothiophene-2-carboxylate ≥98.0%
Methyl-3-bromothiophene-2-carboxylate
Formel:
C₆H₅BrO₂S MW: 221,07 g/mol Kokpunkt: 127°C |
MDL Number:
MFCD00173839 CAS nummer: 26137-08-6 |
Specifikation testresultat
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 97.0 % |
Melting point | 47.0 to 51.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...