Methyl-3-(2-bromophenyl)propionate ≥98.0%
Leverantör: TCI
Synonyms: 3-(2-Bromophenyl)propionic Acid Methyl Ester, Methyl 3-(2-bromophenyl)propanoate
M2587-1G M2587-5G
TCIAM2587-1GEA 365 SEK
TCIAM2587-1G TCIAM2587-5G
Methyl-3-(2-bromophenyl)propionate ≥98.0%
Methyl-3-(2-bromophenyl)propionate
Formel: C₁₀H₁₁BrO₂
Kokpunkt: 87°C
Storage Temperature: Kylskåp
MDL Number: MFCD09953149
CAS nummer: 66191-86-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e103e18082076c2659b402dd46f328723fe50bc&vid=7e103e18082076c2659b402dd46f328723fe50bc&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %