2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
N-(2-Methyl-5-nitrophenyl)-4-(3-pyridyl)-2-pyrimidinamine, 2-(2-Methyl-5-nitroanilino)-4-(3-pyridyl)pyrimidine
M2584-1G
M2584-5G
TCIAM2584-1GEA
295
SEK
TCIAM2584-1G
TCIAM2584-5G
2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine ≥98.0% (by HPLC, titration analysis)
2-(2-Methyl-5-nitrophenylamino)-4-(3-pyridyl)pyrimidine
Formel:
C₁₆H₁₃N₅O₂ MW: 307,313 g/mol |
MDL Number:
MFCD02179269 CAS nummer: 152460-09-8 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 192.0 to 196.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...