3-Methyl-1-butene ∼12,5% ∼1.5 mol/L i THF

Leverantör: TCI EUROPE
Fara

Synonyms: Isoamylene, Isopropylethylene, Isopentene

M2565-100ML M2565-500ML
TCIAM2565-100MLEA 575 SEK
TCIAM2565-100ML TCIAM2565-500ML
3-Methyl-1-butene ∼12,5% ∼1.5 mol/L i THF
3-Methyl-1-butene
Formel: C₅H₁₀
MW: 70,13 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00008937
CAS nummer: 563-45-1
EINECS: 209-249-1
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5060f040928548e8c1ee67802214bc9b15e67f21&vid=5060f040928548e8c1ee67802214bc9b15e67f21&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 12.0 to 14.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR