3-Methyl-1-butene ∼15% ∼2.5 mol/L i diklormetan

Leverantör: TCI EUROPE
Fara

Synonyms: Isoamylene, Isopropylethylene, Isopentene

M2563-100ML M2563-500ML
TCIAM2563-100MLEA 845 SEK
TCIAM2563-100ML TCIAM2563-500ML
3-Methyl-1-butene ∼15% ∼2.5 mol/L i diklormetan
3-Methyl-1-butene
Formel: C₅H₁₀
MW: 70,13 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00008937
CAS nummer: 563-45-1
EINECS: 209-249-1
UN: 1593
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f70de3d887801771db74b3578806902eeaac8551&vid=f70de3d887801771db74b3578806902eeaac8551&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 15.0% to 17.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR