(3,4-Dichlorophenyl)acetic acid methyl ester ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Methyl 3,4-Dichlorophenylacetate, 3,4-Dichlorophenylacetic acid methyll ester, Methyl 2-(3,4-dichlorophenyl)acetate, Methyl (3,4-dichlorophenyl)acetate
M2557-5G
M2557-25G
TCIAM2557-5GEA
895
SEK
TCIAM2557-5G
TCIAM2557-25G
(3,4-Dichlorophenyl)acetic acid methyl ester ≥98.0%
(3,4-Dichlorophenyl)acetic acid methyl ester
Formel:
(C₆H₃Cl₂)CH₂COOCH₃ MW: 219,07 g/mol |
MDL Number:
MFCD00800680 CAS nummer: 6725-44-6 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...