Methyl-2-bromo-3-methylbenzoate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Methyl 2-bromo-3-methylbenzoate
M2546-5G
M2546-25G
TCIAM2546-5GEA
550
SEK
TCIAM2546-5G
TCIAM2546-25G
Methyl-2-bromo-3-methylbenzoate ≥98.0%
Methyl-2-bromo-3-methylbenzoate
Formel:
C₉H₉BrO₂ MW: 229,07 g/mol |
MDL Number:
MFCD00144762 CAS nummer: 131001-86-0 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...