Methyl-5-methyl-2-nitrobenzoate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 6-Nitro-m-toluic acid methyll ester, Methyl 6-Nitro-m-toluate, Methyl 5-methyl-2-nitrobenzoate, 5-Methyl-2-nitrobenzoic acid methyll ester

M2544-5G M2544-25G
TCIAM2544-5GEA 455 SEK
TCIAM2544-5G TCIAM2544-25G
Methyl-5-methyl-2-nitrobenzoate ≥98.0%
Methyl-5-methyl-2-nitrobenzoate
Formel: C₉H₉NO₄
MW: 195,1721 g/mol
MDL Number: MFCD00157275
CAS nummer: 20587-30-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c37554deb5ae6a17612a5da8cb260cd3ea7ff538&vid=c37554deb5ae6a17612a5da8cb260cd3ea7ff538&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR