Methyl-3-bromo-2-methylbenzoate ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Methyl 3-bromo-2-methylbenzoate
M2543-5G M2543-25G
TCIAM2543-5GEA 975 SEK
TCIAM2543-5G TCIAM2543-25G
Methyl-3-bromo-2-methylbenzoate ≥98.0%
Methyl-3-bromo-2-methylbenzoate
Formel: C₉H₉BrO₂
MW: 229,0706 g/mol
Kokpunkt: 140°C
MDL Number: MFCD09031772
CAS nummer: 99548-54-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=784da154424bf55ba6743e962d4764ebb762923b&vid=784da154424bf55ba6743e962d4764ebb762923b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %