(2,4-Dichlorophenyl)acetic acid methyl ester ≥99.0%

Leverantör: TCI EUROPE

Synonyms: Methyl 2,4-Dichlorophenylacetate, Methyl (2,4-dichlorophenyl)acetate, Methyl 2,4-dichlorophenylacetate, 2,4-Dichlorophenylacetic acid methyll ester

M2528-5G M2528-25G
TCIAM2528-5GEA 710 SEK
TCIAM2528-5G TCIAM2528-25G
(2,4-Dichlorophenyl)acetic acid methyl ester ≥99.0%
(2,4-Dichlorophenyl)acetic acid methyl ester
Formel: (C₆H₃Cl₂)CH₂COOCH₃
MW: 219,07 g/mol
Kokpunkt: 153°C
MDL Number: MFCD00018381
CAS nummer: 55954-23-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7b2e12daadac2ae03c0e17b5d686d5b99d3faaa&vid=c7b2e12daadac2ae03c0e17b5d686d5b99d3faaa&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR