Molsidomine 98,0 HPLC, T
Leverantör: TCI
|
Varning
|
Synonyms:
(Ethoxycarbonyl)(3-morpholino-1,2,3-oxadiazol-3-ium-5-yl)amide
M2524-1G
M2524-5G
TCIAM2524-1GEA
830
SEK
TCIAM2524-1G
TCIAM2524-5G
Molsidomine 98,0 HPLC, T
Molsidomine
Formel:
C₉H₁₄N₄O₄ MW: 242.24 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00869301 CAS nummer: 25717-80-0 EINECS: 247-207-4 Merck Index: 14,06234 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | max. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 139.0 to 143.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...