2-Methyl-5-propionylfuran ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
1-(5-Methylfuran-2-yl)propan-1-one
M2507-5G
M2507-25G
TCIAM2507-5GEA
445
SEK
TCIAM2507-5G
TCIAM2507-25G
2-Methyl-5-propionylfuran ≥98.0%
2-Methyl-5-propionylfuran
Formel:
C₈H₁₀O₂ MW: 138,17 g/mol Kokpunkt: 72°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00053142 CAS nummer: 10599-69-6 EINECS: 234-215-8 |
Specifikation testresultat
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...