2-(4-Methyl-5-thiazolyl)ethyl decanoate ≥98.0%

Leverantör: TCI
Varning

Synonyms: Decanoic Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester, 5-(2-Decanoyloxyethyl)-4-methylthiazole, 2-(4-Methylthiazol-5-yl)ethyl decanoate

M2490-5G M2490-25G
TCIAM2490-5GEA 390 SEK
TCIAM2490-5G TCIAM2490-25G
2-(4-Methyl-5-thiazolyl)ethyl decanoate ≥98.0%
2-(4-Methyl-5-thiazolyl)ethyl decanoate
Formel: C₁₆H₂₇NO₂S
MW: 297.46 g/mol
Kokpunkt: 181°C
MDL Number: MFCD09032915
CAS nummer: 101426-31-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7c3de25d09d76afa78068d1e8d285fe4df7930e&vid=d7c3de25d09d76afa78068d1e8d285fe4df7930e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR