2-Methyl-5-oxo-1-cyclopentenyl butyrate ≥98.0%
Leverantör: TCI
Synonyms:
2-Methyl-5-oxocyclopent-1-en-1-yl butyrate, Butyric Acid 2-Methyl-5-oxo-1-cyclopentenyl Ester
M2486-5G
M2486-25G
TCIAM2486-5GEA
895
SEK
TCIAM2486-5G
TCIAM2486-25G
2-Methyl-5-oxo-1-cyclopentenyl butyrate ≥98.0%
2-Methyl-5-oxo-1-cyclopentenyl butyrate
Formel:
C₁₀H₁₄O₃ MW: 182.22 g/mol Kokpunkt: 85°C |
MDL Number:
MFCD00798052 CAS nummer: 68227-51-0 EINECS: 269-363-2 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...