6-Methylnicotinamide ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI
Varning

M2477-5G M2477-25G
TCIAM2477-5GEA 1275 SEK
TCIAM2477-5G TCIAM2477-25G
6-Methylnicotinamide ≥98.0% (by HPLC, titration analysis)
6-Methylnicotinamide
Formel: C₇H₈N₂O
MDL Number: MFCD00006342
CAS nummer: 6960-22-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95fd10d504cc155aa85acf65de3c39627989c812&vid=95fd10d504cc155aa85acf65de3c39627989c812&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 197.0 to 201.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR