Methyl-2-bromoisonicotinate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Methyl 2-bromoisonicotinate

M2461-1G M2461-5G
TCIAM2461-1GEA 655 SEK
TCIAM2461-1G TCIAM2461-5G
Methyl-2-bromoisonicotinate ≥98.0%
Methyl-2-bromoisonicotinate
Formel: C₇H₆BrNO₂
MW: 216,0320 g/mol
MDL Number: MFCD03791265
CAS nummer: 26156-48-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bc45f3cb7e2d03482f486f93df43b5e9a57c502&vid=9bc45f3cb7e2d03482f486f93df43b5e9a57c502&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 34.0 to 38.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR