1-Methyl-1H-tetrazole ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
M2451-1G
M2451-5G
TCIAM2451-1GEA
855
SEK
TCIAM2451-1G
TCIAM2451-5G
1-Methyl-1H-tetrazole ≥98.0%
1-Methyl-1H-tetrazole
Formel:
C₂H₄N₄ MW: 84,08 g/mol Kokpunkt: 81°C |
MDL Number:
MFCD00129972 CAS nummer: 16681-77-9 UN: 1325 ADR: 4.1,III |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 37.0 to 41.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...