Methyl-3-hydroxyisoxazole-5-carboxylate ≥98.0%

Leverantör: TCI
Varning

M2441-1G M2441-5G
TCIAM2441-1GEA 445 SEK
TCIAM2441-1G TCIAM2441-5G
Methyl-3-hydroxyisoxazole-5-carboxylate ≥98.0%
Methyl-3-hydroxyisoxazole-5-carboxylate
Formel: C₅H₅NO₄
MDL Number: MFCD00062870
CAS nummer: 10068-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=801de258b4cbc9c8519e102481ff4eaed4d5a33a&vid=801de258b4cbc9c8519e102481ff4eaed4d5a33a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 162.0 to 166.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR