1-Methylxanthine ≥97.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 3,7-Dihydro-1-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-1-methylpurine, 1-Methyl-1H-purine-2,6(3H,7H)-dione

M2432-50MG M2432-200MG
TCIAM2432-50MGEA 685 SEK
TCIAM2432-50MG TCIAM2432-200MG
1-Methylxanthine ≥97.0% (by HPLC, titration analysis)
1-Methylxanthine
Formel: C₆H₆N₄O₂
MW: 166,14 g/mol
MDL Number: MFCD00005561
CAS nummer: 6136-37-4
EINECS: 228-108-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c165abbe112a2923daaa13cbd5239c847c55b479&vid=c165abbe112a2923daaa13cbd5239c847c55b479&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR